Assay Details
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013047.1 | NP_001013065.1 | 13 - 14 | 1514 | 78 | |
XM_008764483.1 | XP_008762705.1 | 12 - 13 | 1547 | 78 | ||
XM_008764484.1 | XP_008762706.1 | 12 - 13 | 1547 | 78 | ||
GenBank mRNA | BC079400.1 | - | 13 - 14 | 1661 | 78 | |
BC091225.1 | - | 13 - 14 | 1514 | 78 | ||
FQ224358.1 | - | 12 - 13 | 1552 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
