Assay Details
Gene Symbol: Fam71e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145401.1 | NP_001138873.1 | 6 - 7 | 962 | 75 | |
XM_008758921.1 | XP_008757143.1 | 6 - 7 | 1307 | 75 | ||
XM_008758922.1 | XP_008757144.1 | 6 - 7 | 1280 | 75 | ||
GenBank mRNA | BC161877.1 | - | 6 - 7 | 962 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
