Assay Details
Gene Symbol: Fam71e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145401.1 | NP_001138873.1 | 9 - 10 | 2473 | 77 | |
XM_008758921.1 | XP_008757143.1 | 9 - 10 | 2818 | 77 | ||
XM_008758922.1 | XP_008757144.1 | 9 - 10 | 2791 | 77 | ||
GenBank mRNA | BC161877.1 | - | 10 - 11 | 1380 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
