Assay Details
Gene Symbol: Sgpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191811.1 | NP_001178740.1 | 1 - 2 | 84 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
