Assay Details
Gene Symbol: Sgpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191811.1 | NP_001178740.1 | 2 - 3 | 248 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
