Assay Details
Gene Symbol: Myc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012603.2 | NP_036735.2 | 2 - 3 | 1330 | 90 | |
GenBank mRNA | AY679729.1 | - | 2 - 3 | 1167 | 90 | |
AY679730.1 | - | 2 - 3 | 1330 | 90 | ||
BC091699.1 | - | 2 - 3 | 1136 | 7303974 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
