Assay Details
Gene Symbol: Syt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012665.1 | NP_036797.1 | 3 - 4 | 585 | 86 | |
GenBank mRNA | M64488.1 | - | 3 - 4 | 585 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
