Assay Details
Gene Symbol: Icam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012967.1 | NP_037099.1 | 2 - 3 | 364 | 63 | |
GenBank mRNA | BC081837.1 | - | 2 - 3 | 376 | 7122397 | 63 |
D00913.1 | - | 2 - 3 | 364 | 63 | ||
FQ228241.1 | - | 2 - 3 | 384 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
