Assay Details
Gene Symbol: Tnfaip8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014039.1 | NP_001014061.1 | 1 - 2 | 161 | 62 | |
GenBank mRNA | BC079019.1 | - | 1 - 2 | 161 | 7113794 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC079019.1, NM_001014039.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
