Assay Details
Gene Symbol: Noc4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014129.1 | NP_001014151.1 | 4 - 5 | 485 | 104 | |
GenBank mRNA | BC088275.1 | - | 4 - 5 | 485 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
