Assay Details
Gene Symbol: Maob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013198.1 | NP_037330.1 | 9 - 10 | 1084 | 89 | |
GenBank mRNA | BC078886.1 | - | 9 - 10 | 985 | 89 | |
BC089814.1 | - | 9 - 10 | 1049 | 89 | ||
FQ210326.1 | - | 9 - 10 | 1145 | 89 | ||
M23601.1 | - | 9 - 10 | 1084 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
