Assay Details
Gene Symbol: Clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017106.1 | NP_058802.1 | 3 - 4 | 409 | 78 | |
GenBank mRNA | D50497.1 | - | 3 - 4 | 409 | 78 | |
Z56277.1 | - | 3 - 4 | 361 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
