Assay Details
Gene Symbol: Clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017106.1 | NP_058802.1 | 4 - 5 | 606 | 101 | |
GenBank mRNA | D50497.1 | - | 4 - 5 | 606 | 101 | |
Z56277.1 | - | 4 - 5 | 558 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
