Assay Details
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017194.1 | NP_058890.1 | 11 - 12 | 1367 | 57 | |
XM_006246678.2 | XP_006246740.1 | 12 - 13 | 1482 | 57 | ||
XM_008767807.1 | XP_008766029.1 | 12 - 13 | 1468 | 57 | ||
XM_008767808.1 | XP_008766030.1 | 13 - 14 | 2134 | 57 | ||
GenBank mRNA | X13252.1 | - | 11 - 12 | 1364 | 57 | |
X74836.1 | - | 11 - 12 | 1367 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
