Assay Details
Gene Symbol: Padi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017226.1 | NP_058922.1 | 6 - 7 | 715 | 94 | |
GenBank mRNA | J05022.1 | - | 6 - 7 | 715 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
