Assay Details
Gene Symbol: Mapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017347.2 | NP_059043.1 | 6 - 7 | 929 | 107 | |
GenBank mRNA | AF155236.1 | - | 6 - 7 | 905 | 107 | |
M38194.1 | - | 6 - 7 | 866 | 107 | ||
M61177.1 | - | 6 - 7 | 887 | 107 | ||
S46779.1 | - | 6 - 7 | 929 | 107 | ||
U12008.1 | - | 6 - 7 | 866 | 107 | ||
X65198.1 | - | 6 - 7 | 929 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
