Assay Details
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019238.2 | NP_062111.1 | 2 - 3 | 275 | 73 | |
XM_008770746.1 | XP_008768968.1 | 1 - 2 | 2922 | 73 | ||
GenBank mRNA | BC081810.1 | - | 2 - 3 | 275 | 73 | |
FQ213323.1 | - | 2 - 3 | 258 | 73 | ||
M95591.1 | - | 2 - 3 | 293 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
