Assay Details
Gene Symbol: Necab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022302.1 | NP_071638.1 | 4 - 5 | 325 | 101 | |
GenBank mRNA | AF193755.1 | - | 4 - 5 | 325 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
