Assay Details
Gene Symbol: Necab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022302.1 | NP_071638.1 | 6 - 7 | 566 | 89 | |
GenBank mRNA | AF193755.1 | - | 6 - 7 | 566 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
