Assay Details
Gene Symbol: Dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022934.1 | NP_075223.1 | 3 - 4 | 432 | 62 | |
XM_006238099.1 | XP_006238161.1 | 3 - 4 | 530 | 62 | ||
GenBank mRNA | BC062009.1 | - | 3 - 4 | 389 | 62 | |
FQ216722.1 | - | 3 - 4 | 421 | 62 | ||
U53922.1 | - | 3 - 4 | 432 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
