Assay Details
Gene Symbol: Clpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022947.1 | NP_075236.1 | 8 - 9 | 1110 | 98 | |
XM_008759796.1 | XP_008758018.1 | 9 - 10 | 1464 | 98 | ||
GenBank mRNA | AB027570.1 | - | 8 - 9 | 1110 | 98 | |
BC070917.1 | - | 8 - 9 | 1073 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
