Assay Details
Gene Symbol: Gch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024356.1 | NP_077332.1 | 2 - 3 | 557 | 73 | |
GenBank mRNA | FQ220953.1 | - | 2 - 3 | 544 | 73 | |
M58364.1 | - | 2 - 3 | 557 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
