Assay Details
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031085.2 | NP_112347.1 | 13 - 14 | 2091 | 100 | |
XM_008764720.1 | XP_008762942.1 | 13 - 14 | 1973 | 100 | ||
GenBank mRNA | BC061763.1 | - | 14 - 15 | 2422 | 100 | |
BC081782.1 | - | 13 - 14 | 2091 | 100 | ||
X68400.1 | - | 13 - 14 | 1932 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
