Assay Details
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031085.2 | NP_112347.1 | 5 - 6 | 890 | 76 | |
XM_008764720.1 | XP_008762942.1 | 5 - 6 | 772 | 76 | ||
GenBank mRNA | BC061763.1 | - | 6 - 7 | 1221 | 76 | |
BC081782.1 | - | 5 - 6 | 890 | 76 | ||
X68400.1 | - | 5 - 6 | 731 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
