Assay Details
Gene Symbol: Tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031326.1 | NP_112616.1 | 1 - 2 | 204 | 108 | |
GenBank mRNA | AB014089.1 | - | 1 - 2 | 204 | 108 | |
AF377866.1 | - | 1 - 2 | 211 | 108 | ||
AJ312746.1 | - | 1 - 2 | 204 | 108 | ||
BC062022.1 | - | 1 - 2 | 206 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
