Assay Details
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031816.1 | NP_114004.1 | 1 - 2 | 283 | 81 | |
GenBank mRNA | AF090306.1 | - | 1 - 2 | 283 | 81 | |
BC062012.1 | - | 1 - 2 | 192 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF090306.1, BC062012.1, NM_031816.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
