Assay Details
Gene Symbol: Ccnh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052981.2 | NP_443213.2 | 5 - 6 | 826 | 86 | |
GenBank mRNA | AF154914.1 | - | 4 - 5 | 523 | 86 | |
AJ276495.1 | - | 5 - 6 | 826 | 86 | ||
BC059109.1 | - | 4 - 5 | 629 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
