Assay Details
Gene Symbol: Ccnh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052981.2 | NP_443213.2 | 7 - 8 | 1068 | 74 | |
GenBank mRNA | AF154914.1 | - | 6 - 7 | 765 | 74 | |
AJ276495.1 | - | 7 - 8 | 1068 | 74 | ||
BC059109.1 | - | 6 - 7 | 871 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
