Assay Details
Gene Symbol: Glrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053296.1 | NP_445748.1 | 6 - 7 | 753 | 109 | |
XM_006232506.2 | XP_006232568.1 | 7 - 8 | 899 | 109 | ||
GenBank mRNA | AJ310839.1 | - | 6 - 7 | 753 | 109 | |
FQ212388.1 | - | 7 - 8 | 904 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
