Assay Details
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053427.1 | NP_445879.1 | 2 - 3 | 800 | 92 | |
XM_006229265.1 | XP_006229327.1 | 2 - 3 | 329 | 92 | ||
GenBank mRNA | AF271235.1 | - | 2 - 3 | 800 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
