Assay Details
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053486.1 | NP_445938.1 | 6 - 7 | 2201 | 110 | |
GenBank mRNA | AF083330.1 | - | 6 - 7 | 2171 | 110 | |
AJ223599.1 | - | 6 - 7 | 2201 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
