Assay Details
Gene Symbol: Chek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053677.1 | NP_446129.1 | 11 - 12 | 1358 | 68 | |
XM_006249518.2 | XP_006249580.1 | 12 - 13 | 1472 | 68 | ||
XM_006249519.2 | XP_006249581.1 | 11 - 12 | 1351 | 68 | ||
XM_006249520.2 | XP_006249582.1 | 11 - 12 | 1408 | 68 | ||
XM_006249521.2 | XP_006249583.1 | 11 - 12 | 1292 | 68 | ||
XM_008769308.1 | XP_008767530.1 | 11 - 12 | 1351 | 68 | ||
GenBank mRNA | AF134054.1 | - | 11 - 12 | 1358 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
