Assay Details
Gene Symbol: Chek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053677.1 | NP_446129.1 | 2 - 3 | 425 | 66 | |
XM_006249518.2 | XP_006249580.1 | 3 - 4 | 539 | 66 | ||
XM_006249519.2 | XP_006249581.1 | 2 - 3 | 418 | 66 | ||
XM_006249520.2 | XP_006249582.1 | 2 - 3 | 475 | 66 | ||
XM_006249521.2 | XP_006249583.1 | 2 - 3 | 359 | 66 | ||
XM_008769308.1 | XP_008767530.1 | 2 - 3 | 418 | 66 | ||
GenBank mRNA | AF134054.1 | - | 2 - 3 | 425 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
