Assay Details
Gene Symbol: Ankh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053714.1 | NP_446166.1 | 4 - 5 | 817 | 75 | |
GenBank mRNA | AF393241.1 | - | 4 - 5 | 817 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
