Assay Details
Gene Symbol: Scarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054001.2 | NP_446453.1 | 11 - 12 | 1680 | 83 | |
GenBank mRNA | BC061853.1 | - | 11 - 12 | 1680 | 5621697 | 83 |
D10587.1 | - | 11 - 12 | 1619 | 83 | ||
M68965.1 | - | 11 - 12 | 1628 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P533959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168721 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P533959 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P533959 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P533959 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P533959 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701440355 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70064 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95602 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78553 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94678 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701440355 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10771626 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771624 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771634 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771632 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771633 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771629 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771623 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771627 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771628 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771630 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771625 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771631 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10771622 | - | - |
Rat Genome 230 2.0 Array | 1368379_at | - | - |
Rat Genome 230 2.0 Array | 1395327_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371506_at | - | - |
Rat Genome 230 2.0 Array | 1368379_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371506_PM_at | - | - |
Rat Genome 230 2.0 Array | 1395327_at | - | - |
Affymetrix HT Rat Focus Array | 1368379_at | - | - |
Affymetrix HT Rat Focus Array | 1368379_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400001926.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001923.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001928.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001927.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001918.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001917.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001925.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001922.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001919.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001921.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001920.rn.1 | TC1400000249.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001924.rn.1 | TC1400000249.rn.1 | RTA TC |
Rat Expression Set 230 | 1368379_PM_at | - | - |
Rat Expression Set 230 | 1368379_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771622 | - | - |
Rat Genome U34 Set | D10587_g_at | - | - |
Rat Genome U34 Set | D10587_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361889 | - | - |
RatRef-12 Expression BeadChip | ILMN_48587 | - | - |
