Assay Details
Gene Symbol: Chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057184.1 | NP_476532.1 | 3 - 4 | 440 | 86 | |
GenBank mRNA | L08227.1 | - | 3 - 4 | 440 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
