Assay Details
Gene Symbol: Chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057184.1 | NP_476532.1 | 5 - 6 | 1532 | 83 | |
GenBank mRNA | L08227.1 | - | 5 - 6 | 1532 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
