Assay Details
Gene Symbol: Syt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138835.1 | NP_620190.1 | 3 - 4 | 454 | 99 | |
XM_008760071.1 | XP_008758293.1 | 3 - 4 | 382 | 99 | ||
XM_008760072.1 | XP_008758294.1 | 4 - 5 | 428 | 99 | ||
GenBank mRNA | U71294.1 | - | 3 - 4 | 454 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
