Assay Details
Gene Symbol: Podxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138848.1 | NP_620203.1 | 7 - 8 | 1400 | 74 | |
XM_008762758.1 | XP_008760980.1 | 7 - 8 | 1560 | 74 | ||
GenBank mRNA | AB020726.1 | - | 7 - 8 | 1473 | 74 | |
AF109393.1 | - | 7 - 8 | 1400 | 74 | ||
BC070886.1 | - | 7 - 8 | 1470 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
