Assay Details
Gene Symbol: Grasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138894.1 | NP_620249.1 | 2 - 3 | 350 | 89 | |
GenBank mRNA | AF374272.1 | - | 2 - 3 | 350 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
