Assay Details
Gene Symbol: Slc36a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139339.1 | NP_647555.1 | 6 - 7 | 848 | 64 | |
GenBank mRNA | AF512430.1 | - | 6 - 7 | 848 | 64 | |
BC166414.1 | - | 6 - 7 | 782 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
