Assay Details
Gene Symbol: Bcl2l11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171989.1 | NP_741986.1 | 1 - 2 | 122 | 96 | |
XM_006234987.2 | XP_006235049.2 | 2 - 3 | 503 | 96 | ||
GenBank mRNA | AF065432.1 | - | 1 - 2 | 122 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
