Assay Details
Gene Symbol: Arhgap39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173122.3 | NP_775145.2 | 11 - 12 | 3363 | 84 | |
XM_006241866.2 | XP_006241928.1 | 10 - 11 | 3318 | 84 | ||
XM_006241867.2 | XP_006241929.1 | 10 - 11 | 3308 | 84 | ||
GenBank mRNA | CB578147.1 | - | 3 - 4 | 460 | 84 | |
X53232.1 | - | 1 - 2 | 106 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the X53232.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
