Assay Details
Gene Symbol: Arhgap39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173122.3 | NP_775145.2 | 4 - 5 | 819 | 93 | |
XM_006241867.2 | XP_006241929.1 | 4 - 5 | 857 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
