Assay Details
Gene Symbol: Arhgap39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173122.3 | NP_775145.2 | 6 - 7 | 2741 | 97 | |
XM_006241866.2 | XP_006241928.1 | 5 - 6 | 2696 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
