Assay Details
Gene Symbol: Arhgap39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173122.3 | NP_775145.2 | 8 - 9 | 2908 | 65 | |
XM_006241866.2 | XP_006241928.1 | 7 - 8 | 2863 | 65 | ||
XM_006241867.2 | XP_006241929.1 | 7 - 8 | 2853 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
