Assay Details
Gene Symbol: Arhgap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004242.1 | NP_001004242.1 | 8 - 9 | 788 | 66 | |
GenBank mRNA | BC079089.1 | - | 8 - 9 | 788 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
