Assay Details
Gene Symbol: Ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007705.1 | NP_001007706.1 | 3 - 4 | 365 | 113 | |
XM_006240475.2 | XP_006240537.1 | 3 - 4 | 421 | 113 | ||
XM_006240476.2 | XP_006240538.1 | 3 - 4 | 421 | 113 | ||
GenBank mRNA | BC081966.1 | - | 3 - 4 | 365 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
