Assay Details
Gene Symbol: Tph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173839.2 | NP_776211.1 | 2 - 3 | 349 | 109 | |
XM_006241352.2 | XP_006241414.1 | 2 - 3 | 402 | 109 | ||
XM_008765357.1 | XP_008763579.1 | 2 - 3 | 402 | 109 | ||
GenBank mRNA | AY098915.1 | - | 2 - 3 | 349 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
