Assay Details
Gene Symbol: Ppm1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175755.2 | NP_786931.1 | 3 - 4 | 440 | 61 | |
XM_006248674.1 | XP_006248736.1 | 3 - 4 | 545 | 61 | ||
XM_008768853.1 | XP_008767075.1 | 3 - 4 | 491 | 61 | ||
XM_008768854.1 | XP_008767076.1 | 3 - 4 | 527 | 61 | ||
XM_008768855.1 | XP_008767077.1 | 3 - 4 | 545 | 61 | ||
GenBank mRNA | AB023634.1 | - | 3 - 4 | 440 | 61 | |
BC166854.1 | - | 3 - 4 | 545 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
